Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Surgical and Dissection Equipment
▸
Scalpel Blades
▸
BiopBlade™ Sterile Blades
Actual product may vary
Integra Miltex
BiopBlade™ Sterile Blades
Flexible design allows the blade to be positioned at any angle for the intended procedure
Have Questions?
Catalog Number
Quantity
FIS12-460-391
50/Box
Catalog number
FIS12-460-391
Price (SGD)
362.00
Each
Add to cart
Quantity:
50/Box
Price (SGD)
362.00
Each
Add to cart
For skin/tissue saucerization and shave biopsy
Disposable, one-time use only
Sterile, 5-year shelf life
Rear finger guard
Specifications
Product Type
Surgical Blade
For Use With (Application)
For Skin/Tissue Saucerization, Shave Biopsy, Dermatology, Podiatry and Plastic Surgery
Disposable
Single-use
Sterility
Sterile
Quantity
50/Box
Shelf Life
5 Years