Instrument Cleaning Brushes
Actual product may vary
Instrument Cleaning Brushes
Instrument Cleaning Brushes
Integra Miltex
Instrument Cleaning Brushes
Reusable and autoclavable
Have Questions?
Catalog NumberMaterial
FIS12-460-389Nylon
Catalog number FIS12-460-389
Price (SGD)
67.00
Each
Material:
Nylon
Price (SGD)
67.00
Each
  • Brush is 18.4cm (7.25in.) long
Nylon bristle brushes (12460389)
  • Used for most instruments including delicate ones
  • Steel bristle brushes (12460390)
    • Better for rasps, files and burrs
    Specifications
    Product TypeInstrument Cleaning Brush
    Color-codedNo
    AutoclavableAutoclavable
    For Use With (Application)For most instruments including delicate ones
    Length (English)7.25 in.
    Length (Metric)18.4 cm
    MaterialNylon
    Unit SizeEach