Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Surgical and Dissection Equipment
▸
Scalpel Handles
▸
Scalpel Handles
Actual product may vary
Integra Miltex
Scalpel Handles
Have Questions?
Catalog Number
Size
Blade Material
Length (English)
FIS12-460-109
# 3
Stainless Steel
5 in.
Catalog number
FIS12-460-109
Price (SGD)
79.00
Each
Add to cart
Size:
# 3
Blade Material:
Stainless Steel
Length (English):
5 in.
Price (SGD)
79.00
Each
Add to cart
Available in stainless steel and autoclavable plastic
Reusable
Specifications
Blade Material
Stainless Steel
Product Type
Scalpel Handle
Size
# 3
Length (English)
5 in.
Length (Metric)
12.7 cm
For Use With (Equipment)
# 10 to # 15C Blade
Disposable
Reusable
Graduated
Yes