Immersion Oil
Immersion Oil
Actual product may vary
Cargille
Immersion Oil
Greatly increases light-gathering power with high-power objectives
Have Questions?
Change viewbuttonViewtableView
Catalog NumberViscosityDensityQuantityDescription
FIS12-370A150 cSt0.923g/cm34 oz.Type A Immersion Oil
FIS12-368A150 cSt0.923g/cm31 oz.Type A Immersion Oil
FIS12-368B150 cSt0.923g/cm316 oz.Type A Immersion Oil
FIS12-369B1250 cSt0.923g/cm316 oz.Type B Immersion Oil
FIS12-370B1250 cSt0.923g/cm34 oz.Type B Immersion Oil
FIS12-369A1250 cSt0.923g/cm31 oz.Type B Immersion Oil
Catalog number FIS12-370A
Price (SGD)
-
Viscosity:
150 cSt
Density:
0.923g/cm3
Quantity:
4 oz.
Description:
Type A Immersion Oil
Greatly increases light-gathering power with high-power objectives
  • Formulated to give refractive index of 1.5150 ± 0.0002 for D line at 23°C (73°F)
  • Nondrying, so index will not change when exposed to air
  • Contains no polychlorinated biphenyl (PCB) compounds
  • Conforms to ISO 8036/1 specifications
  • Standard for all oil-immersion microscopy
  • Type A oil has low viscosity and low fluorescence
  • Type B is a high-viscosity oil used in biomedical microscopy
  • Type NVH is a high-viscosity oil that meets requirements for horizontal, inverted and inclined instruments and projection equipment
  • Type FF oil is virtually nonfluorescent
  • Type DF is a very low-fluorescence oil for fluorescence microscopy with very high resolution
Specifications
Viscosity150 cSt
Certifications/ComplianceISO 8036/1
Density0.923g/cm3
DescriptionType A Immersion Oil
For Use With (Application)Increases light-gathering power with high-power objectives
Quantity4 oz.
Refractive Index1.5150 ±0.0002
Unit SizeEach