Check Order Status
Blue Polythene Gauntlet Gloves
Actual product may vary
Fisherbrand
Blue Polythene Gauntlet Gloves
Shermond™ Disposable Blue Polythene Arm Length Gauntlets are food safe and intended for single use.
Have Questions?
Catalog number
FIS12-20
Price (SGD)
-
Fully disposable arm-length gauntlets
Food safe polythene
Ideal for general purpose use including food handling and also veterinary applications
Ambidextrous fit
Powder free and non-sterile
Supplied in 50 packs of 50 gloves
Not suitable for clinical applications
Specifications
Color
Blue
Exterior Finish
No Texture
Hand
Ambidextrous
Latex Free
Yes
Material
Polyethylene
For Use With (Application)
General Purpose use Including Food Hygiene
Glove Cuff Style
Gauntlet