Check Order Status
RNase-Free Disposable Pellet Pestles
Actual product may vary
Fisherbrand
RNase-Free Disposable Pellet Pestles
Individually wrapped to ensure purity
Have Questions?
Change view
Catalog Number
Includes
Capacity (English)
FIS12-141-364
Without tube
0.05 oz.
FIS12-141-368
With tube
0.05 oz.
FKS13236679
Without tube
0.05 oz.
3 Options
Catalog number
FIS12-141-364
Price (SGD)
127.00
CS
Add to cart
Includes:
Without tube
Capacity (English):
0.05 oz.
Price (SGD)
127.00
CS
Add to cart
Individually wrapped to ensure purity
RNase, DNase and Pyrogen-free
Volume: 1.5mL
Specifications
Autoclavable
Autoclavable
Color
Blue
Capacity (English)
0.05 oz.
Capacity (Metric)
1.5 mL
Material
Polypropylene
Product Type
Pellet Pestle
Disposable
Single-use
Length (English)
2.75 in.
Dnase Free
Yes
Includes
Without tube
Length (Metric)
70mm
Capacity
0.05 oz., 1.5mL