Rtx™-1701 Capillary Columns
Rtx™-1701 Capillary Columns
Actual product may vary
Bausch and Lomb
Rtx™-1701 Capillary Columns
General-Purpose Columns for Alcohols, Oxygenates, PCB Congeners (e.g., Aroclor Mixes), Pesticides, and Fragrance Compounds
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric) InnerFilm Thickness
FIS12-05015 m0.25 mm1 μm
Catalog number FIS12-050
Price (SGD)
-
Length (Metric):
15 m
Diameter (Metric) Inner:
0.25 mm
Film Thickness:
1 μm
  • Mix of cyano and phenyl functional groups increase polarity
  • Offers a different elution order relative to less polar Rtx™-1 or Rtx™-5 columns
  • Ideal for confirmation analysis in combination with an Rtx™-35 or Rtx™-5 column
  • Polymer is fully characterized to ensure long-term reproducibility, column-to-column consistency, and low bleed
  • For Alcohols, Oxygenates, PCB Congeners (e.g., Aroclor Mixes), Pesticides, and Fragrance Compounds
Specifications
Functionality/Interaction TypeCyano and Phenyl
Diameter (Metric) Inner0.25 mm
Stationary PhaseRtx™-1701
USP TypeG46
For Use With (Application)Alcohols, Oxygenates, PCB Congeners (e.g., Aroclor Mixes), Pesticides, and Fragrance Compounds
Particle Size1 μm
Film Thickness1 μm
Length (Metric)15 m
MaterialFused Silica
PolarityMidpolar
CoatingFused Silica
Quantity1
Unit SizeEA