Hastings Triplet Adjustable Focus Measuring Magnifying Glass
Actual product may vary
Bausch and Lomb
Hastings Triplet Adjustable Focus Measuring Magnifying Glass
Provides a flat image without distortion
Have Questions?
Catalog NumberMagnificationDiopterLength (English) Focal
FIS12-032-0310x40 diopter1 in.
Catalog number FIS12-032-03
Price (SGD)
-
Magnification:
10x
Diopter:
40 diopter
Length (English) Focal:
1 in.
  • Power viewing area presents a clean, sharp, crisp image with incorporated Bausch & Lomb precision optics
  • Lens adjusts for individual focus
  • Transparent body allows natural light illumination on the magnified area
  • Protective case keeps the magnifier safe, scratch free in all work environments
  • Scale sold separately (12-037D)
Specifications
Diopter40 diopter
For Use With (Application)To provide a flat image without distortion
IncludesProtective case and storage pouch for case and lens
Length (English) Focal1 in.
Length (Metric) Focal2.54 cm
Magnification10x
MaterialGlass
Product TypeMagnifier, Hastings Triplet Measuring
Unit SizeEach