Baume Hydrometers
Baume Hydrometers
Actual product may vary
Baume Hydrometers
Baume heavy scale for working with liquids heavier than water.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCertifications/ComplianceIncrementsLength (Metric)Baume Range
FIS11571FC3-point Certification, NIST, ASTM E100, ASTM E1260.1°295 mm50 to 61°
FIS11-571AC3-point Certification, NIST, ASTM E100, ASTM E1260.1°310 mm0 to 12°
FIS11-571CC3-point Certification, NIST, ASTM E100, ASTM E1260.1°310 mm20 to 31°
FIS11-571BC3-point Certification, NIST, ASTM E100, ASTM E1260.1°305 mm9 to 21°
FIS11-571DC3-point Certification, NIST, ASTM E100, ASTM E1260.1°305 mm29 to 41°
FIS11-571EC3-point Certification, NIST, ASTM E100, ASTM E1260.1°290 mm40 to 51°
FIS11571EC3-point Certification, NIST, ASTM E100, ASTM E1260.1°290 mm40 to 51°
FIS11-580AC3-point Certification, NIST, ASTM E100, ASTM E1260.5°280 mm0 to 30°
FIS11-580DWithout 3-point certification0.5°300 mm35 to 70°
FIS11-580CC3-point Certification, NIST, ASTM E100, ASTM E126285 mm0 to 70°
FIS11-580EWithout 3-point certification300 mm50 to 100°
FIS11-571BWithout 3-point certification0.1°305 mm9 to 21°
FIS11-571DWithout 3-point certification0.1°305 mm29 to 41°
FIS11-571AWithout 3-point certification0.1°310 mm0 to 12°
FIS11-571CWithout 3-point certification0.1°310 mm19 to 31°
FIS11-571EWithout 3-point certification0.1°290 mm40 to 51°
FIS11-571FWithout 3-point certification0.1°295 mm49 to 61°
FIS11-580CWithout 3-point certification285 mm0 to 70°
FIS11-571GWithout 3-point certification0.1°295 mm60 to 70°
FIS11-580BWithout 3-point certification0.5°305 mm0 to 50°
FIS11-580AWithout 3-point certification0.5°280 mm0 to 30°
Catalog number FIS11571FC
Price (SGD)
780.00
EA
Add to cart
Certifications/Compliance:
3-point Certification, NIST, ASTM E100, ASTM E126
Increments:
0.1°
Length (Metric):
295 mm
Baume Range:
50 to 61°
Price (SGD)
780.00
EA
Add to cart
Baume heavy scale for working with liquids heavier than water.
  • Permanently marked with individual serial numbers
Specifications
TypeBaume Hydrometer
Increments0.1°
Certifications/Compliance3-point Certification, NIST, ASTM E100, ASTM E126
Length (Metric)295 mm
MaterialSoda Lime Glass
Baume Range50 to 61°
For Use With (Application)Baume 'Heavy' Scale For Liquids Heavier than Water
Unit SizeEA