Bibulous Paper for Drying Slides
Bibulous Paper for Drying Slides
Actual product may vary
Bibulous Paper for Drying Slides
Highly absorbent white fiber paper recommended for drying microscope slides and other small lab glass items.
Have Questions?
Catalog NumberQuantity
FIS11-99810,800 Sheets
Catalog number FIS11-998
Price (SGD)
1,924.00
PK
Add to cart
Quantity:
10,800 Sheets
Price (SGD)
1,924.00
PK
Add to cart
Absorbent white paper
Highly absorbent white fiber paper recommended for drying microscope slides and other small lab glass items.
  • Lint and dust free to clean without leaving streaks
  • Each sheet measures 6 x 4 in. (15 x 10 cm)
  • Each book contains 50 sheets neatly bound with heavy paper covers
Available in Pack (12 books, 600 sheets) or Case (18 packs, 10,800 sheets).
Specifications
TypeBibulous Paper
Dimensions (L x W)6 x 4 in. (15 x 10 cm)
For Use With (Equipment)Drying slides
Length (English)6 in.
Length (Metric)15 cm
Quantity10,800 Sheets
Width (English)4 in.
Width (Metric)10 cm
Unit SizePK