Lens Paper
Lens Paper
Actual product may vary
Fisherbrand
Lens Paper
For cleaning glass lenses
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Quantity
FIS11-9964 x 6 in.50 Sheets/Book, 12 Books/Pk.
FIS11-9956 x 8 in. sheets100 Sheets/Book, 12 Books/Pk.
Catalog number FIS11-996
Price (SGD)
53.00
PK
Add to cart
Dimensions (L x W):
4 x 6 in.
Quantity:
50 Sheets/Book, 12 Books/Pk.
Price (SGD)
53.00
PK
Add to cart
For cleaning glass lenses
Booklet
  • Never scratches glass lenses, objectives, or other delicate surfaces
  • Doesn't become harsh; doesn't readily collect dust or grease
  • Virtually lint-free sheets are glued and bound in a booklet for easy removal
Dispenser Box
  • Same paper as above, but packaged in a convenient dispenser box for fast, single-sheet removal individual sheets measure 5L × 7 in.W (13 × 18cm)
Specifications
Product TypeLens Cleaning Paper
Dimensions (L x W)4 x 6 in.
For Use With (Application)For cleaning Glass lenses
Quantity50 Sheets/Book, 12 Books/Pk.
Unit SizePK