Germicidal UV Sterilizing Lamps
Actual product may vary
Spectroline
Germicidal UV Sterilizing Lamps
Unfiltered 254nm shortwave (UV-C) lamps provide high-intensity, wide area UV sterilization coverage
Have Questions?
Catalog NumberFor Use With (Equipment)
FIS11-992-1035W Short wave Spectroline Single Tube
Catalog number FIS11-992-103
Price (SGD)
1,671.00
Each
For Use With (Equipment):
5W Short wave Spectroline Single Tube
Price (SGD)
1,671.00
Each
  • For use wherever disease may be transmitted by air currents
  • Choose from 15w single-tube or 15w dual-tube models
  • All units have enameled sheet metal or anodized aluminum housing with mounting brackets
Specifications
Light SourceGermicidal UV-C
Wattage15 W
Dimensions (L x W x H)18.75 x 4.5 x 3.5 in. (476.2 x 114.3 x 88.9 mm)
For Use With (Equipment)5W Short wave Spectroline Single Tube
Height (English)3.5 in.
Height (Metric)88.9 mm
Length (English)18.75 in.
Length (Metric)476.2 mm
Packaging Quantity1 Ea.
Width (English)4.5 in.
Width (Metric)114.3 mm
Product TypeGermicidal UV Sterilizing Lamp