AutoclavableAutoclavable
Closure TypeNone
ColorBlue
DisposableSingle-use
For Use With (Application)General hospital use
MaterialNon-woven Fabric
SterilityNon-sterile
Dimensions (L x W)12 x 12 in. (30.4 x 30.4 cm)
Length (English)12 in.
Width (English)12 in.
Product TypeSteri-Wrap I
Length (Metric)30.4 cm
Width (Metric)30.4 cm
Unit SizeCase