Steri-Wrap™ III Cellulose Based Sterilization Wrap
Actual product may vary
Steri-Wrap™ III Cellulose Based Sterilization Wrap
Steri-Wrap™ III Cellulose Based Sterilization Wrap
Propper
Steri-Wrap™ III Cellulose Based Sterilization Wrap
Steri-Wrap™ III is a cellulose based heavy weight wrap suitable for your heaviest instruments in steam and ethylene oxide sterilization cycles. Efficient microbial barrier, high-level of protection against fluids and particles. Highly permeable meaning no wet packs.
Have Questions?
Catalog NumberLength (English)Width (English)
FIS11-890-118 in.18 in.
Catalog number FIS11-890-1
Price (SGD)
1,041.00
Case
Length (English):
18 in.
Width (English):
18 in.
Price (SGD)
1,041.00
Case
  • Steri-Wrap™ is cellulose based sterilization wrap available in three grades for keeping packages safe and secure during the sterilization process.
  • Steri-Wrap™ III is a heavy weight wrap suitable for your heaviest instrument packages.
  • Steri-Wrap™ is designed to be virtually lint free with tightly anchored fibers.
  • Wrap is reversible and features excellent liquid repellency.
  • Steri-Wrap™ can be used in steam or ethylene oxide sterilization.
  • Variety of strength of fabric to meet your needs.
Specifications
AutoclavableAutoclavable
Closure TypeNone
ColorBlue
DisposableSingle-use
For Use With (Application)OR and CSR wrapping
MaterialNon-woven Fabric
SterilityNon-sterile
No. per Case400/Cs.
Dimensions (L x W)18 x 18 in. (45.7 x 45.7 cm)
Length (English)18 in.
Width (English)18 in.
Product TypeSteri-Wrap III
Length (Metric)45.7 cm
Width (Metric)45.7 cm
Unit SizeCase