Blue Autoclavable Tape
Actual product may vary
Propper
Blue Autoclavable Tape
Blue Autoclavable Tape provides a means of identifying steam-sterilized packs while ensuring their security during sterilization.
Have Questions?
Catalog NumberDimensions (L x W)
FIS11-889-560 yd. x 1 in.
Catalog number FIS11-889-5
Price (SGD)
32.00
Each
Dimensions (L x W):
60 yd. x 1 in.
Price (SGD)
32.00
Each
  • Specially designed with an aggressive adhesive backing to stick securely to all disposable wraps
  • High-contrast, white, steam-sensitive stripes turn dark when subjected to steam autoclaving temperatures
Specifications
AutoclavableAutoclavable
Adhesive TypeRubber
CoatingSynthetic Resin
ColorWhite, Blue
FormatTape
MaterialCrepe Paper
Product TypeSteam Autoclave Tape
SterilityNon-sterile
For Use With (Application)Different types of sterilization packs, to distinguish processed and unprocessed items
Quantity1 Ea.
Dimensions (L x W)60 yd. x 1 in.
Length (English)180 ft.
Length (Metric)54.9 m
Width (English)1 in.
Width (Metric)25.4 mm
Diameter (English)3 in.
Diameter (Metric)7.62 cm
Unit SizeEach