White Autoclave Indicator Tape
White Autoclave Indicator Tape
Actual product may vary
Propper
White Autoclave Indicator Tape
Color change eliminates the guesswork
Have Questions?
Catalog NumberWidth (English)Width (Metric)Length (English)Length (Metric)
FIS11-889-121 in.25 mm180 ft.55 m
Catalog number FIS11-889-12
Price (SGD)
-
Width (English):
1 in.
Width (Metric):
25 mm
Length (English):
180 ft.
Length (Metric):
55 m
Convenient color change eliminates the usual guesswork.
  • For use in steam sterilization applications
  • No need for interpretation—sensitive ink on white stripes turns dark to indicate that pack has been processed
  • Specially formulated universal adhesive provides secure bonding with fabric and nonwoven wraps, paper, metal, and glass
  • Tape removes easily without any traces of residue
Specifications
AutoclavableAutoclavable
Adhesive TypePressure-sensitive
ColorWhite
FormatTape
MaterialCrepe Paper
For Use With (Application)Steam Sterilization
Quantity36/Cs.
Length (English)180 ft.
Length (Metric)55 m
Width (English)1 in.
Width (Metric)25 mm
Unit SizeEA