Autoclave Indicator Tapes
Actual product may vary
Propper
Autoclave Indicator Tapes
Ideal to secure different types of sterilization packs
Have Questions?
Catalog NumberdimColor
FIS11-889-10
Catalog number FIS11-889-10
Price (SGD)
-
  • Pressure-sensitive self-adhesive tape
  • Material: crepe-embossed paper
  • Bleading resistant to permanent markers after sterilization
For labeling products for steam and ethylene oxide sterilization
Specifications
Product TypeAutoclave Indicator Tape
For Use With (Equipment)Autoclaves
MaterialCrepe Paper
Length (English)60 yd.
Width (English)0.5 in.
Length (Metric)54.9 m
Adhesive TypeSelf-adhesive
AutoclavableAutoclavable
For Use With (Application)Steam or Ethyline Oxide sterilization
Quantity1 Ea.
Unit SizeEach