GHS Labeled Right-to-Know Safety-Vented Wash Bottles
Actual product may vary
GHS Labeled Right-to-Know Safety-Vented Wash Bottles
GHS Labeled Right-to-Know Safety-Vented Wash Bottles
GHS Labeled Right-to-Know Safety-Vented Wash Bottles
Zeiss
GHS Labeled Right-to-Know Safety-Vented Wash Bottles
Unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips and allows dispensing of wash bottle in upright or inverted position.
Have Questions?
Catalog NumberChemical Label
FIS11-865-163Ethanol
Catalog number FIS11-865-163
Price (SGD)
-
Chemical Label:
Ethanol
  • Pre-printed bottles with chemical name and formula, GHS pictogram and signal word; National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Translucent LDPE bottles, Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
  • Wide mouth 53mm (2 in.) for easy refilling
  • Color-coded polypropylene closures aid in quick and easy identification
  • Assortment bag includes one each: Acetone, Isopropanol, Bleach and Ethanol
    Bottle with Label: TOLUENE
    • Red LDPE bottle
    • Does not include vented valve
    Bottle with Label: DEIONIZED H2O, DISTILLED H20 and WATER
    • Does not indicate hazardous chemical and does not have/require GHS pictogram or signal word
    Bottle with Label: METHYL ETHYL KETONE
    • Does not include vented valve
    Compliance: Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA’s HazCom 2012 requirements – as pertains to “Workplace Labels”.
Specifications
Capacity (Metric)500 mL
ColorNatural
MaterialLDPE
Quantity3/Bag
Closure ColorNatural
MouthWide
Chemical LabelEthanol
Closure MaterialPolypropylene
Closure Size53 mm
Closure TypeCap