Self-Adhesive Labels
Self-Adhesive Labels
Actual product may vary
Self-Adhesive Labels
Use these white labels on a variety of surfaces. Fisherbrand™ Self-Adhesive Labels are removable and pressure sensitive. Ideal for a wide range of labeling applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorWidth (English)Length (English)Quantity
FIS11-863EWhite0.75 in.1.5 in.500/Sheet
FIS11-850BWhite, Red1.5 in.2 in.540/Box
FIS11-850CWhite, Red1.25 in.1.62 in.720/Box
FIS11-863DWhite0.75 in.1 in.1000/Sheet
Catalog number FIS11-863E
Price (SGD)
-
Color:
White
Width (English):
0.75 in.
Length (English):
1.5 in.
Quantity:
500/Sheet
Use these white labels on a variety of surfaces. Fisherbrand™ Self-Adhesive Labels are removable and pressure sensitive. Ideal for a wide range of labeling applications.
  • Self Adhesive Labels
  • Stick To Most Surfaces Without Moistening
  • Easily Marked With Pencil, Ballpoint Pen, or felt-tip Marker
Specifications
Adhesive TypeSelf-adhesive
ColorWhite
FormatLabel
MaterialPaper
Product TypeSelf Adhesive Label
For Use With (Application)General purpose
Quantity500/Sheet
ShapeRectangle
Dimensions (LxW)1.5 in. x 0.75 in.
Length (English)1.5 in.
Length (Metric)3.8 cm
Width (English)0.75 in.
Unit SizePK