Plain Form Petroleum Hydrometers
Plain Form Petroleum Hydrometers
Actual product may vary
Plain Form Petroleum Hydrometers
API Plain Form Petroleum Hydrometers are available in regular and ASTM™ recommended forms.
Have Questions?
Catalog NumberCertifications/ComplianceAPI Range
FIS11-603SC3-point Certification, NIST, Cited in ASTM E100-14 Table 1, ASTM E12649 to 61° ASTM 6H-62
Catalog number FIS11-603SC
Price (SGD)
-
Certifications/Compliance:
3-point Certification, NIST, Cited in ASTM E100-14 Table 1, ASTM E126
API Range:
49 to 61° ASTM 6H-62
  • Permanently marked with individual serial numbers
  • Without thermometer in body
  • Approximate length: 152mm
  • Use Fisherbrand Cylinder 08-530E; 140mL liquid volume
  • Temperature of standardization: 60°F
Specifications
Increments0.2°
Certifications/Compliance3-point Certification, NIST, Cited in ASTM E100-14 Table 1, ASTM E126
Length (Metric)152 mm
For Use With (Equipment)175mL Borosilicate Glass cylinder
MaterialSoda Lime Glass
For Use With (Application)Petroleum products
Temperature of Standardization60°F
API Range49 to 61° ASTM 6H-62
Unit SizeEA