ASTM™ Hydrometers, Plain Form
ASTM™ Hydrometers, Plain Form
Actual product may vary
ASTM™ Hydrometers, Plain Form
API hydrometers are available in regular and ASTM™ recommended forms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberASTM NumberCertifications/Compliance
FIS11-603E7HNIST, Cited in ASTM E100-14 Table 1, ASTM E126
FIS11-603C5HNIST, Cited in ASTM E100-14 Table 1, ASTM E126
FIS11-603B4HNIST, Cited in ASTM E100-14 Table 1, ASTM E126
Catalog number FIS11-603E
Price (SGD)
51.00
EA
Add to cart
ASTM Number:
7H
Certifications/Compliance:
NIST, Cited in ASTM E100-14 Table 1, ASTM E126
Price (SGD)
51.00
EA
Add to cart
These hydrometers are manufactured to meet or exceed ASTM™ E 100 specifications which are accepted by NIST™ and U.S. Bureau of Mines.
  • Permanently marked with individual serial numbers
  • Without thermometer in body
  • Approximate length: 330mm
  • Temperature of standardization: 60°F
Specifications
ASTM Number7H
Increments0.1°
Certifications/ComplianceNIST, Cited in ASTM E100-14 Table 1, ASTM E126
Length (Metric)330 mm
MaterialSoda Lime Glass
Temperature of Standardization60°F
Unit SizeEA