1x1 image pixel for data collection
Alcohol Hydrometers
Alcohol Hydrometers
Actual product may vary
Alcohol Hydrometers
Used by distillers, law enforcement and taxing agencies to measure the alcoholic content of beverages.
Have Questions?
Catalog NumberCertifications/Compliance
FIS11-590NIST, ASTM E100, ASTM E126
Catalog number FIS11-590
Price (SGD)
54.00
EA
Estimated availability date 17-Sep-2025
Add to cart
Certifications/Compliance:
NIST, ASTM E100, ASTM E126
Price (SGD)
54.00
EA
Add to cart
Used by distillers, law enforcement and taxing agencies to measure the alcoholic content of beverages.
Calibrated on two scales: Tralle (0% by volume alcohol) and proof (2× percentage of alcohol).
  • Tralle scale, 0 to 100% graduated in 1% divisions
  • Proof scale, 0 to 200 in two proof divisions
  • Permanently marked with individual serial numbers
  • Approximate liquid volume, 250mL
Specifications
Increments1%
Certifications/ComplianceNIST, ASTM E100, ASTM E126
Length (Metric)300 mm
MaterialSoda Lime Glass
For Use With (Application)Distillers, law enforcement and taxing agencies to measure the Alcohol content of beverages
Volume (Metric)250 mL
Twaddell Range0 to 100°
Unit SizeEA
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link