Twaddell Hydrometers
Product Image
Twaddell Hydrometers
For use with liquids heavier than water such as milk, sulfuric acid and tanning solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCertifications/ComplianceLength (Metric)Twaddell Range
FIS11-585AC3-point Certification, NIST, ASTM E100, ASTM E126300 mm0 to 26°
FIS11-585AWithout 3-pt certification290 mm0 to 26°
Catalog number FIS11-585AC
Price (SGD)
-
Certifications/Compliance:
3-point Certification, NIST, ASTM E100, ASTM E126
Length (Metric):
300 mm
Twaddell Range:
0 to 26°
  • Permanently marked with individual serial numbers
Specifications
Increments0.5°
Certifications/Compliance3-point Certification, NIST, ASTM E100, ASTM E126
Length (Metric)300 mm
For Use With (Equipment)300mL Borosilicate Glass cylinder
For Use With (Application)Liquids heavier than Water such as Milk, Sulfuric Acid, Tanning solutions
Twaddell Range0 to 26°
Unit SizeEA