Replacement Hydrometers for FBA11582 Precision Specific Gravity Hydrometer Set
Replacement Hydrometers for FBA11582 Precision Specific Gravity Hydrometer Set
Actual product may vary
Replacement Hydrometers for FBA11582 Precision Specific Gravity Hydrometer Set
Replacement Hydrometers for the FBA11582 Hydrometer Set.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCertifications/ComplianceIncrementsSpecific Gravity Range
FIS11-583HWithout 3-pt Certification0.0021.8 to 2
FIS11-583AWithout 3-pt Certification0.0010.700 to 0.810
FIS11-583AC3-point Certification0.0010.700 to 0.810
FIS11-583BWithout 3-pt Certification0.0010.800 to 0.910
FIS11-583BC3-point Certification0.0010.800 to 0.910
FIS11-583EWithout 3-pt Certification0.0021.200 to 1.420
FIS11-583CWithout 3-pt Certification0.0010.900 to 1.010
FIS11-583DWithout 3-pt Certification0.0021.000 to 1.220
FIS11-583DC3-point Certification0.0021.000 to 1.220
FIS11-583EC3-point Certification0.0021.200 to 1.420
FIS11-583FWithout 3-pt Certification0.0021.400 to 1.620
FIS11-583FC3-point Certification0.0021.400 to 1.620
FIS11-583GWithout 3-pt Certification0.0021.600 to 1.820
FIS11-583GC3-point Certification0.0021.600 to 1.820
Catalog number FIS11-583H
Price (SGD)
56.00
EA
Add to cart
Certifications/Compliance:
Without 3-pt Certification
Increments:
0.002
Specific Gravity Range:
1.8 to 2
Price (SGD)
56.00
EA
Add to cart
Replacement Hydrometers for the FBA11582 Hydrometer Set.
Replacement Hydrometer for FBA11582 Hydrometer Set.
Specifications
Increments0.002
Certifications/ComplianceWithout 3-pt Certification
Length (Metric)300 mm
For Use With (Equipment)FBA11582 Hydrometer Set
MaterialSoda Lime Glass
For Use With (Application)Specific Gravity Measurement
Specific Gravity Range1.8 to 2
Unit SizeEA