Specific Gravity Hydrometers
Specific Gravity Hydrometers
Actual product may vary
Specific Gravity Hydrometers
Measure true specific gravity of liquids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCertifications/ComplianceIncrementsLength (Metric)Specific Gravity Range
FIS11-522DC3-point Certification, ASTM E100, ASTM E1260.02165 mm1.000 to 2.000
FIS11-510AASTM E100, ASTM E1260.001310 mm0.700 to 0.810
FIS11-510CASTM E100, ASTM E1260.001305 mm0.900 to 1.000
FIS11-510CC3-point Certification, ASTM E100, ASTM E1260.001305 mm0.900 to 1.000
FIS11-510DASTM E100, ASTM E1260.005305 mm0.650 to 1.000
FIS11-512AASTM E100, ASTM E1260.002180 mm0.700 to 0.800
FIS11-510DC3-point Certification, ASTM E100, ASTM E1260.005305 mm0.650 to 1.000
FIS11-512BASTM E100, ASTM E1260.002170 mm0.800 to 0.900
FIS11-512CC3-point Certification, ASTM E100, ASTM E1260.002180 mm0.900 to 1.000
FIS11-512BC3-point Certification, ASTM E100, ASTM E1260.002170 mm0.800 to 0.900
FIS11-512DASTM E100, ASTM E1260.01165 mm0.650 to 1.000
FIS11-520AASTM E100, ASTM E1260.002305 mm1.000 to 1.220
FIS11-520AC3-point Certification, ASTM E100, ASTM E1260.002305 mm1.000 to 1.220
FIS11-520BASTM E100, ASTM E1260.002275 mm1.200 to 1.420
FIS11-520CC3-point Certification, ASTM E100, ASTM E1260.002285 mm1.400 to 1.620
FIS11-520EASTM E100, ASTM E1260.01305 mm1.000 to 2.000
FIS11-520CASTM E100, ASTM E1260.002285 mm1.400 to 1.620
FIS11-520BC3-point Certification, ASTM E100, ASTM E1260.002275 mm1.200 to 1.420
FIS11-520EC3-point Certification, ASTM E100, ASTM E1260.01305 mm1.000 to 2.000
FIS11-520FASTM E100, ASTM E1260.005290 mm1.000 to 1.600
FIS11-522AASTM E100, ASTM E1260.005185 mm1.000 to 1.225
FIS11-522AC3-point Certification, ASTM E100, ASTM E1260.005185 mm1.000 to 1.225
FIS11-522BASTM E100, ASTM E1260.005160 mm1.200 to 1.425
FIS11-522CASTM E100, ASTM E1260.005160 mm1.400 to 1.625
FIS11-522BC3-point Certification, ASTM E100, ASTM E1260.005160 mm1.200 to 1.425
FIS11-522DASTM E100, ASTM E1260.02165 mm1.000 to 2.000
Catalog number FIS11-522DC
Price (SGD)
802.00
EA
Add to cart
Certifications/Compliance:
3-point Certification, ASTM E100, ASTM E126
Increments:
0.02
Length (Metric):
165 mm
Specific Gravity Range:
1.000 to 2.000
Price (SGD)
802.00
EA
Add to cart
Measure true specific gravity of liquids.
  • Chemical-resistant bulbs and stems
  • Streamlined to avoid trapping air bubbles
  • With directional device that stops the instrument with the scale facing the user for more convenient, more accurate readings
  • Double scales can be read without turning the hydrometer.
Specifications
TypeSpecific Gravity Hydrometer
Increments0.02
Certifications/Compliance3-point Certification, ASTM E100, ASTM E126
Length (Metric)165 mm
For Use With (Equipment)300mL Borosilicate Glass cylinder
MaterialSoda Lime Glass
For Use With (Application)Measure true specific gravity of liquids heavier than Water
Specific Gravity Range1.000 to 2.000
Volume (Metric)140 mL
Unit SizeEA