Standard Weight Hot Mill Gloves
Actual product may vary
Wells Lamont
Standard Weight Hot Mill Gloves
Standard Weight Hot Mill Gloves are constructed of standard weight, 24 oz. terrycloth and protect from temperatures up to 300°F.
Have Questions?
Catalog NumberSize
FIS11-462-6ALarge
Catalog number FIS11-462-6A
Price (SGD)
118.00
Dozen
Size:
Large
Price (SGD)
118.00
Dozen
  • Constructed of standard weight, 24 oz. terrycloth
  • Gloves protect from temperatures up to 300°F
  • Straight thumb
  • Nap-out
Specifications
Coating MaterialNap-out
ColorNatural
MaterialTerrycloth
Product TypeHot Mill Gloves
SizeLarge
Temperature (English) Max.300°F
Temperature (Metric) Max149°C
For Use With (Application)Steel Processing, Metal Handling
Glove Cuff StyleSafety
SeriesY6243
Thumb StyleStraight
Yarn Weight24 oz.
Unit SizeDozen