Specifications
Capacity (Metric)8.5 L
Dimensions (L x W x H)12.75 x 10.5 x 4.25 in.
ColorBlue
Height (Metric)10.8 cm
Length (Metric)32.4 cm
MaterialPolypropylene
TypeMultipurpose Tray
Width (Metric)26.7 cm
Length (English)12.75 in.
Height (English)4.25 in.
ShapeRectangular
Width (English)10.5 in.
Capacity (English)9 qt.