4-Isopropylphenoxyacetic acid, 98+%
4-Isopropylphenoxyacetic acid, 98+%
4-Isopropylphenoxyacetic acid, 98+%
Bel-Art
4-Isopropylphenoxyacetic acid, 98+%
CAS: 1643-16-9 | C11H14O3 | 194.23 g/mol
Have Questions?
Catalog NumberQuantity
FIS11-394-45425 g
Catalog number FIS11-394-454
Price (SGD)
137.00
Each
Quantity:
25 g
Request bulk or custom format
Price (SGD)
137.00
Each
Specifications
Chemical Name or Material4-Isopropylphenoxyacetic acid
CAS1643-16-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c