Cryo-Gloves™ Blue Waterproof Elbow Length Cryogenic Gloves
Actual product may vary
Tempshield
Cryo-Gloves™ Blue Waterproof Elbow Length Cryogenic Gloves
A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).
Have Questions?
Catalog NumberSize
FIS11-394-331X-Large
Catalog number FIS11-394-331
Price (SGD)
687.00
Pair
Size:
X-Large
Price (SGD)
687.00
Pair
A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).
  • Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
  • Thin, seamless 100% waterproof liner provides protection from spills and splashes
  • State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
  • High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
  • Ideal for applications where function is important and safety is critical
  • Not intended for immersion in liquid nitrogen or other cryogenic liquids
Specifications
Certifications/Compliance (Europe)EN 420, EN 511
DescriptionElbow Length
ColorBlue
HandSpecific
Product TypeWaterproof Cryogenic Gloves
SizeX-Large
EN388 StandardYes
For Use With (Application)Dispensing/Transferring Cryogenic Liquids, Removing Samples from Cryogenic Liquids, and Open Cryogenic Containers with the Chance of Exposure to Liquids/Splashes
SeriesCryo-Gloves™
Temperature (English) Min-320°F
Temperature (Metric) Min.-196°C
Length (English)17.3 to 19.8 in.
Length (Metric)44 to 50 cm
Unit SizePair