1-Decylboronic acid, 98%
1-Decylboronic acid, 98%
1-Decylboronic acid, 98%
Tempshield
1-Decylboronic acid, 98%
CAS: 24464-63-9 | C10H23BO2 | 186.102 g/mol
Have Questions?
Catalog NumberQuantity
FIS11-394-3261 g
Catalog number FIS11-394-326
Price (SGD)
557.00
Pair
Quantity:
1 g
Request bulk or custom format
Price (SGD)
557.00
Pair
Specifications
Chemical Name or Material1-Decylboronic acid
CAS24464-63-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c