Bench-Top Biohazard Bag Holder
Actual product may vary
Bel-Art
Bench-Top Biohazard Bag Holder
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
FIS11-394-310Biohazard Bag Holder Kit
FIS11-394-303Bag Holder Cover
FIS01-815-2Biohazard Bag Holder
Catalog number FIS11-394-310
Price (SGD)
183.00
Each
Product Type:
Biohazard Bag Holder Kit
Price (SGD)
183.00
Each
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.
  • Rectangular steel wire frame to hold one polyethylene bag of dimensions 8.5 x 11 in. (22 x 28cm)
  • Available with epoxy coat in orange color
  • Comes with snug fitting polypropylene plastic cover to hold bag securely in place
  • Reusable and allows cleaning by hand or in automated dish washers
  • Autoclavable
  • Also used with Clavies Autoclavable Bags H13182-0812
Specifications
AutoclavableAutoclavable
ColorPink
DisposableSingle-use
For Use With (Application)Perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use
For Use With (Equipment)01-828A, 01-828-031, 01-830A, 01-826-1, 01-827A, 01-829A, 01-815-1 22 x 28cm biohazard bag, Clavies™ autoclavable bags (01-815-6)
IncludesComplete Kit: Bag Holder, Cover and Bags (100pk)
MaterialPolypropylene
Width (English)5 in.
Product TypeBiohazard Bag Holder Kit
Width (Metric)12.7 cm
Unit SizeEach