S-Thiouronium chloride, polymer-supported, 0.8-1.0 mmol/g on Merrifield resin, Thermo Scientific™
Have Questions?
Catalog Number
Quantity
FIS11-394-306
1 g
Catalog number FIS11-394-306
Price (SGD)
562.00
Pair
Quantity:
1 g
Request bulk or custom format
Price (SGD)
562.00
Pair
Specifications
Chemical Name or MaterialS-Thiouronium chloride
Name Notepolymer-supported, 0.8 to 1.0mmol/g on Merrifield resin
Recommended StorageAmbient temperatures
TSCANo
MDL NumberMFCD03458600
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.