Cryo-Gloves™ Blue Mid-Arm Length Cryogenic Gloves
Actual product may vary
Cryo-Gloves™ Blue Mid-Arm Length Cryogenic Gloves
State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS11-394-305Medium
FIS11-394-200Large
Catalog number FIS11-394-305
Price (SGD)
452.00
Pair
Size:
Medium
Price (SGD)
452.00
Pair
State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).
  • Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
  • State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
  • High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
  • Ideal for applications where function is important and safety is critical
  • Not intended for immersion in liquid nitrogen or other cryogenic liquids
Specifications
Certifications/Compliance (Europe)EN 420, EN 511
DescriptionMid-Arm Length
ColorBlue
HandSpecific
Product TypeCryogenic Gloves
SizeMedium
EN388 StandardYes
For Use With (Application)Dry Ice Handling, Low/Ultra-Low Temperature Freezers, and Closed Cryogenic Systems
SeriesCryo-Gloves™
Temperature (English) Min-320°F
Temperature (Metric) Min.-196°C
Length (English)13.3 to 15.5 in.
Length (Metric)33.5 to 39.5 cm
Unit SizePair