Clavies™ Heat-Resistant Biohazard Autoclave Gloves
Actual product may vary
Clavies™ Heat-Resistant Biohazard Autoclave Gloves
Clavies™ Heat-Resistant Biohazard Autoclave Gloves
Bel-Art
Clavies™ Heat-Resistant Biohazard Autoclave Gloves
Soft, pliant, cotton terrycloth gloves are ideal for handling hot objects out of the autoclave or oven and protect from temperatures up to 450°F (232°C).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English) Cuff
FIS11-394-2995 in.
FIS11-394-29811 in.
Catalog number FIS11-394-299
Price (SGD)
114.00
Pair
Length (English) Cuff:
5 in.
Price (SGD)
114.00
Pair
Soft, pliant, cotton terrycloth gloves are ideal for handling hot objects out of the autoclave or oven and protect from temperatures up to 450°F (232°C).
  • Ideal for removing hot objects from an autoclave or oven
  • Heat-resistant up to 232°C
  • Replaces hazardous asbestos gloves
  • Extended gauntlet cuff available for additional forearm protection
  • Gloves are not waterproof and should not be used to handle dry ice
  • Machine wash, air dry, or use the Lab-Aire™ II Glove Holder (F18931-0000) to dry on any Lab-Aire™ dryer
Specifications
Certifications/ComplianceCE
Accelerator FreeYes
ApplicationsLaboratory, Industrial
Coating MaterialUncoated
ColorOrange
Latex FreeYes
MaterialTerrycloth
Product TypeAutoclave Gloves
SizeUniversal
StyleFull-hand Mitt
Temperature (English) Max.450°F
Temperature (Metric) Max232°C
For Use With (Application)Handling Hot Objects
Glove Cuff StyleGauntlet
SeriesH13201
Length (English)13 in.
Length (English) Cuff5 in.
Length (Metric)33 cm
Length (Metric) Cuff13 cm
Unit SizePair