Zetex™ 100 Series Heat Resistant Gloves
Actual product may vary
Newtex
Zetex™ 100 Series Heat Resistant Gloves
Gloves are constructed from Zetex™ Fiberglass Fabric for protection from temperatures up to 1000°F for extended periods of time.
Have Questions?
Catalog NumberLength (English)
FIS11-392-5114 in.
Catalog number FIS11-392-51
Price (SGD)
254.00
Pair
Length (English):
14 in.
Price (SGD)
254.00
Pair
  • Premium wool liner
  • Double stitched and reinforced
  • Inorganic fibers will not produce hazardous outgassing
Specifications
Coating MaterialUncoated
ColorWhite
MaterialZetex Fiberglass Fabric
SizeUniversal
Temperature (English) Max.1000°F (Extended), 1300°F (Intermittent)
Temperature (Metric) Max540°C (Extended), 700°C (Intermittent)
For Use With (Application)Kiln/Furnace Operations, Heat Processing Facilities, and High Temperature Industrial Environments
Glove Cuff StyleOpen
Liner MaterialWool
SeriesZetex™ 100
Length (English)14 in.
Length (Metric)35.5 cm
Unit SizePair