Model 818 Basic Glove Box
Actual product may vary
Plas Labs
Model 818 Basic Glove Box
The Plas Lab Model 818 Basic Glove Box is a basic model that fits general laboratory isolation applications, but is easily modified for more specialized uses.
Have Questions?
Catalog NumberDepth (Metric)Depth (Metric) Interior
FIS11-389-120119 cm97 cm
Catalog number FIS11-389-120
Price (SGD)
40,956.00
Each
Depth (Metric):
119 cm
Depth (Metric) Interior:
97 cm
Price (SGD)
40,956.00
Each
Compact, portable and lightweight glove boxes for isolating sensitive research studies from a hostile exterior environment.
  • Optically clear, one piece acrylic top
  • Coved, easy-to-clean corners with no hidden dark spots
  • Double-layered, closed cellular neoprene gasketing system
  • Ambidextrous, white Hypalon™ gloves provide superior chemical and UV resistance; stainless-steel 0-rings for gloves
  • Adjustable vacuum gauge on transfer chamber
  • Four ground key cock valves for purging
  • White leveling tray for transferring liquids through the transfer chamber
  • All clamps fully adjustable to compensate for wear
  • Transparent transfer chamber, 12L x 11 in. l.D.
11-389
  • Matched die-molded, bright white bottom
  • 11-389-120
    • Multistation model
    • Two sets of glove ports
    Specifications
    Product TypeGlove Box
    Certifications/ComplianceUL, CA
    Depth (English)47 in.
    Depth (English) Interior38 in.
    Depth (Metric)119 cm
    Depth (Metric) Interior97 cm
    For Use With (Application)General laboratory isolation applications in pharmaceutical, biotech, animal science, waste management, industrial/contract research, cleanroom facilities, microelectronics assembly, chemical labs and plasma welding
    Height (English)42 in.
    Height (English) Interior26 in.
    Height (Metric)107 cm
    Volume (Metric)1158 L
    Width (English)76 in.
    Width (English) Interior60 in.
    Width (Metric)193 cm
    Width (Metric) Interior152 cm
    Unit SizeEach