Hypalon™ Glove Box Gloves for Anaerobic Chamber
Actual product may vary
Plas Labs
Hypalon™ Glove Box Gloves for Anaerobic Chamber
One-piece gloves with 8 in. hand size and 10 in. cuff.
Have Questions?
Catalog NumberDiameter (Metric) Cuff
FIS11-389-125 cm
Catalog number FIS11-389-1
Price (SGD)
1,995.00
Each
Diameter (Metric) Cuff:
25 cm
Price (SGD)
1,995.00
Each
For use with Plas-Labs Anaerobic Chamber (11-391-106), Plas-Labs Basic Glove Box (11-389), and Plas-Labs Analytical Balance Chamber (11-389-6)
Specifications
ColorWhite
HandAmbidextrous
MaterialCSM
Size8
Model800-GH
Diameter (English) Cuff10 in.
Diameter (Metric) Cuff25 cm
Unit SizeEach