Model 830-CP Compact Glove Box
Actual product may vary
Plas Labs
Model 830-CP Compact Glove Box
Plas Lab Model 830-CP Compact Glove Box is designed specifically for use when weighing hazardous materials that must be contained.
Have Questions?
Catalog NumberDepth (English)Depth (English) InteriorVolume (English)
FIS11-386-14131 in.29 in.17.6 cu. ft.
Catalog number FIS11-386-141
Price (SGD)
34,689.00
Each
Depth (English):
31 in.
Depth (English) Interior:
29 in.
Volume (English):
17.6 cu. ft.
Price (SGD)
34,689.00
Each
Offer cleanliness, safety, portability and a draft-free atmosphere for the maximum effectiveness of your balance.
  • Models include access doors; some models also include vents
  • Top section is removable for easy installation of equipment
  • Main chamber holds two purging gas valves
  • Models offer a white leveling tray for easy transferring of liquids, white ambidextrous Hypalon™ gloves for superior chemical and UV resistance, a pressure relief valve that minimizes glove “fight back,” and replaceable HEPA and organic vapor canister filters
  • All clamps are stainless steel and completely adjustable
Specifications
Product TypeGlove Box
Filter TypeHEPA, Organic Vapor
Certifications/ComplianceUL, CSA, CE
Depth (English)31 in.
Depth (English) Interior29 in.
Depth (Metric)78.74 cm
Depth (Metric) Interior73.66 cm
For Use With (Application)Determining the dry weight or moisture content of aqueous solutions, adhesives, cereals, toxic chemicals, pharmaceuticals, paper, plastics and light radioactive materials
Height (English)34 in.
Height (English) Interior30 in.
Height (Metric)86 cm
Volume (English)17.6 cu. ft.
Warranty1 Year
Width (English)63 in.
Width (English) Interior35 in.
Width (Metric)160 cm
Width (Metric) Interior91.44 cm
Unit SizeEach