Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Specialty Lab Glassware
▸
Beads and Boiling Aids
▸
Economical Solid Glass Beads for Distillation Columns
Actual product may vary
Walter Stern
Economical Solid Glass Beads for Distillation Columns
Used for mixing, boiling, or packing
Have Questions?
Change view
Catalog Number
Diameter (Metric)
FIS11-312C
5 mm
FIS11-312B
4 mm
FIS11-312A
3 mm
3 Options
Catalog number
FIS11-312C
Price (SGD)
34.00
Pack
Add to cart
Diameter (Metric):
5 mm
Price (SGD)
34.00
Pack
Add to cart
Used for mixing, boiling, or packing
Spherical Soda-lime glass
Chemical resistant
Specifications
Type
Glass Bead
Material
Glass
Diameter (Metric)
5 mm
For Use With (Equipment)
Distillation Columns