Economical Solid Glass Beads for Distillation Columns
Actual product may vary
Walter Stern
Economical Solid Glass Beads for Distillation Columns
Used for mixing, boiling, or packing
Have Questions?
Catalog NumberDiameter (Metric)
FIS11-312B4 mm
Catalog number FIS11-312B
Price (SGD)
37.00
Pack
Diameter (Metric):
4 mm
Price (SGD)
37.00
Pack
Used for mixing, boiling, or packing
  • Spherical Soda-lime glass
  • Chemical resistant
Specifications
TypeGlass Bead
MaterialGlass
Diameter (Metric)4 mm
For Use With (Equipment)Distillation Columns
Unit SizePack