Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Specialty Lab Glassware
▸
Beads and Boiling Aids
▸
Perforated Glass Beads
Actual product may vary
Walter Stern
Perforated Glass Beads
Spread boiling area
Have Questions?
Catalog Number
Diameter (Metric)
FIS11-311A
4 mm
Catalog number
FIS11-311A
Price (SGD)
79.00
Pack
Add to cart
Diameter (Metric):
4 mm
Price (SGD)
79.00
Pack
Add to cart
Soda-lime glass
Packed in 16 oz. (454g) bottles
Specifications
Material
Soda Lime Glass
Diameter (Metric)
4 mm
Quantity
454 g