PYREX™ Gas Dispersion Tubes With Fritted Cylinders
PYREX™ Gas Dispersion Tubes With Fritted Cylinders
Actual product may vary
PYREX™ Gas Dispersion Tubes With Fritted Cylinders
Used to disperse or scrub gas in liquids.
Have Questions?
Catalog NumberPore SizePorosityIncludes
FIS11-138A170 to 220 μmExtra CoarseExtra coarse fritted cylinder
Catalog number FIS11-138A
Price (SGD)
183.00
EA
Add to cart
Pore Size:
170 to 220 μm
Porosity:
Extra Coarse
Includes:
Extra coarse fritted cylinder
Price (SGD)
183.00
EA
Add to cart
These immersion tubes have a 30 mm diameter medium porosity (10 μm to 15 μm) fritted disc and are particularly useful for drawing off the supernatant liquid from a precipitate. They are made from Code 7740 borosilicate glass. Any precipitate which might be disturbed will be held on the filter. Solid chemicals, made in a beaker or flask, can be filtered and washed without being transferred.
Specifications
TypePyrex Gas Dispersion Tube
IncludesExtra coarse fritted cylinder
Pore Size170 to 220 μm
PorosityExtra Coarse
Diameter (English) Stem0.31 in.
Diameter (Metric) Stem8 mm
Length (English)9.8 in.
Length (Metric)250 mm
For Use With (Application)Used to disperse or scrub gas in liquids
For Use With (Equipment)Test tubes or flasks
Unit SizeEA