EBD Series Exam Glove Box Holder
Actual product may vary
EBD Series Exam Glove Box Holder
EBD Series Exam Glove Box Holder
S-Curve
EBD Series Exam Glove Box Holder
Dispensers are constructed of 1/8 in. clear PETG, with one large compartment to keep multiple boxes of exam gloves neat and organized.
Have Questions?
Catalog NumberCapacityDimensions (D x W x H) Interior
FIS11-127-47C3 Boxes3.5 x 10 x 15 in.
Catalog number FIS11-127-47C
Price (SGD)
183.00
Case
Capacity:
3 Boxes
Dimensions (D x W x H) Interior:
3.5 x 10 x 15 in.
Price (SGD)
183.00
Case
  • Constructed of 1/8 in. clear PETG material
  • Keyholes on the back for wall mounting with included hardware
  • Fits boxes with a maximum size of 10 x 3.5 in.
  • Flame polished edges for a smooth finish
Specifications
Product TypeGlove Box Holder
ColorClear
Dimensions (D x W x H) Interior3.5 x 10 x 15 in.
HoldsExam Glove Boxes
MaterialPETG
MountingKeyhole Slots
No. of Compartments1
Product StyleTop Loading
Capacity3 Boxes
IncludesMounting hardware
Thickness (English)0.125 in.
Unit SizeCase