Shandon™ Instrument Case
Shandon™ Instrument Case
Actual product may vary
Thermo Scientific
Shandon™ Instrument Case
Epredia™ Shandon Instrument Case is available as a complete kit or case only.
Have Questions?
Catalog NumberDescription
FIS1001382Case without instruments
Catalog number FIS1001382
Price (SGD)
-
Description:
Case without instruments
Case dimensions:
  • 12.75 x 9.5 x 2.5 in. (32.4 x 24.1 x 6.4cm)
Instruments Available in Varying Sizes and Styles:
  • Dissecting scissors
  • Iris scissors
  • Liston bone shears
  • Rib shears
  • Plain bone shears
  • Liston bone-cutting forceps
  • Councilman's chisel
  • Tissue thumb forcep
  • Broad point dressing thumb forceps
  • Cartilage thumb forceps
  • Halstead's hemostatic mosquito forceps
  • Dura strip forceps
  • Virchow skull breaker
  • Bone mallet
  • Post mortem hammer with hook
  • Grooved needle director
Specifications
Product TypeInstrument Case
DescriptionCase without instruments
Unit SizeEA