Gas Cylinder Support
Gas Cylinder Support
Actual product may vary
Gas Cylinder Support
Keeps cylinders upright and safe
Have Questions?
Catalog number FIS10-595Q
Price (SGD)
-
Keeps cylinders upright and safe
  • Nylon strap securely holds 4.5 to 11 in. diameter (11.4 to 28cm) cylinders upright
  • Can be attached to benches up to 2.25 in. thick (5.7cm), with at least 1 in. (2.5cm) overhang
Specifications
Holds4.5 to 11 in. dia. cylinders upright
Includes47 in. long nylon strap
Product TypeGas Cylinder Support
For Use With (Equipment)Gas cylinder
Unit SizeEA