Corner-Stripping Blades
Actual product may vary
Corner-Stripping Blades
Corner-Stripping Blades
Corner-Stripping Blades
Corner-Stripping Blades
Corner-Stripping Blades
Thermo Scientific
Corner-Stripping Blades
Finger-friendly™ blade cuts materials effectively and is safe to touch
Have Questions?
Catalog NumberColor
FIS10-532White
Catalog number FIS10-532
Price (SGD)
507.00
Case
Color:
White
Price (SGD)
507.00
Case
  • Compatible with Slice craft knives, scalpel, and Precision Knife
  • Compatible handles: 10548, 10589, 10580, 10568
  • Safer than traditional blades
  • Finger-friendly™ edge cuts materials effectively
  • Lasts up to 11.2x longer than steel blades
  • Fewer blade changes = fewer injuries
  • Reduces injuries, lowers costs
  • Non-sparking, non-conductive, non-magnetic
  • Chemically inert, never rusts
  • Oil and lubricant free
  • Does not require sharps box disposal
  • 100% recyclable
  • Blade is safe up to 1600°C
  • Pointed tip initiates cut with difficult-to-penetrate materials
  • Center point reaches corners for easy cutting and trimming
  • 4 single-edge blades per pack
Specifications
Product TypeCorner Stripping
Length (English)1.29 in.
Length (Metric)32.8 mm
MaterialZirconium Oxide
ColorWhite
For Use With (Application)Safety Cutting
Includes4 single-edge blades per pack
No. per Pack4 Pk.
Weight (English)0.03 oz.
Weight (Metric)0.75 g
Width (English)0.24 in.
Width (Metric)6.15 mm
Unit SizeCase