RTX™-50 Capillary Columns: Standard
RTX™-50 Capillary Columns: Standard
Actual product may vary
Thermo Scientific
RTX™-50 Capillary Columns: Standard
Similar to DB-17, DB-17ht, HP-17, HP-50, SPB-50, SP-2250; ideal confirmation column for many environmental applications, including EPA Methods 604, 8060, and 608
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric) InnerFilm Thickness
FIS10-52430 m0.32 mm0.25 μm
Catalog number FIS10-524
Price (SGD)
1,067.00
CS
Add to cart
Length (Metric):
30 m
Diameter (Metric) Inner:
0.32 mm
Film Thickness:
0.25 μm
Price (SGD)
1,067.00
CS
Add to cart
Columns only.
  • Crossbond™ 50% methyl/50% phenyl polysiloxane
  • Midpolarity
  • Optimized film thickness produces excellent separation of chlorinated pesticides and meets all resolution criteria for CLP pesticides method
  • Crossbond technology provides low bleed, long life, and excellent thermal stability
  • Increased phenyl content offers unique polarity
  • Not adversely affected by column rinsing
Specifications
Diameter (Metric) Inner0.32 mm
Stationary PhaseRtx™-50
USP TypeG3
For Use With (Application)Pesticides, Herbicides, Rosin Acids, Phthalate Esters, and Sterols Analysis
Temperature Range40°C to 300/320°C
Film Thickness0.25 μm
Length (Metric)30 m
MaterialFused Silica
PolarityMidpolar
CoatingFused Silica
Quantity1/Pk.
Unit SizeCS