Kappa Goat anti-Mouse, FITC, Polyclonal, Southern Biotech™
Actual product may vary
Kappa Goat anti-Mouse, FITC, Polyclonal, Southern Biotech™
Kappa Goat anti-Mouse, FITC, Polyclonal, Southern Biotech™
Kappa Goat anti-Mouse, FITC, Polyclonal, Southern Biotech™
Kappa Goat anti-Mouse, FITC, Polyclonal, Southern Biotech™
Goat Polyclonal Secondary Antibody
Have Questions?
Catalog NumberQuantity
FIS10-500-21.0 mg
Catalog number FIS10-500-2
Price (SGD)
70.00
Pack
Quantity:
1.0 mg
Price (SGD)
70.00
Pack
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
AntigenKappa
ApplicationsFlow Cytometry, ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
ClassificationPolyclonal
Concentration1.0mg/mL
ConjugateFITC
FormulationSupplied in PBS/NaN3
Host SpeciesGoat
ImmunogenMouse κ light chains
Purification MethodAffinity Purified
Quantity1.0 mg
Regulatory StatusRUO
Primary or SecondarySecondary
Target SpeciesMouse
IsotypeIgG
Unit SizePack
Contents & Storage
Store at 2-8°C; Protect from light