Search Fisher Scientific
| Catalog Number | Capacity (Metric) |
|---|---|
| FIS10-462-863 | 500 mL |
| FIS10-462-859 | 200 mL |
| FIS10-462-856 | 100 mL |
| FIS10-462-857 | 10 mL |
| FIS10-462-860 | 250 mL |
| FIS10-462-864 | 50 mL |
| FIS10-462-861 | 25 mL |
The strength of the flasks have been significantly increased through machine-blown bodies, which are sealed to heavy-tubing necks. The graduation line is sharp and permanent; large white block letters make the inscription easy to read. Supplied with PTFE color-coded, keyhole stoppers.