Check Order Status
Classic Glass Thistle Tube Funnels
Actual product may vary
Classic Glass Thistle Tube Funnels
Designed with a thistle top.
Have Questions?
Catalog Number
Length (Metric) Stem
FIS10-455B
300mm
Catalog number
FIS10-455B
Price (SGD)
21.00
EA
Add to cart
Length (Metric) Stem:
300mm
Price (SGD)
21.00
EA
Add to cart
Top diameter: 45mm
Stem dia.: 6mm
Borosilicate glass
Specifications
Diameter (Metric)Top
45mm
Diameter (Metric) Stem
6mm
Material
Borosilicate Glass
Length (Metric) Stem
300mm
Product Type
Thistle Tube Funnel